MilliporeSigma
  • Tetramethylenedisulfotetramine contaminated milk powder induced status epilepticus in two siblings and two dogs.
Tetramethylenedisulfotetramine contaminated milk powder induced status epilepticus in two siblings and two dogs.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet (2009-10-23)
Utcharee Intusoma, Vorasith Sornsrivichai
ABSTRACT
A cluster of patients with tetramethylenedisulfotetramine (TETS) intoxication was reported in Thailand. Two siblings, a-six-month-old boy and a-four-year-old girl, and their domestic dogs presented with status epilepticus within 10 minutes after ingesting milk prepared from the same tin container of milk powder. Although the cases showed normal neurodevelopment at one-year follow-up, physicians should be informed of this lethal neurotoxic agent, especially in an era of terroristic activity.
Product Description
Supelco
Tetramethylenedisulfotetramine solution, 100 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.