MilliporeSigma
  • Solution NMR mapping of water-accessible residues in the transmembrane beta-barrel of OmpX.
Solution NMR mapping of water-accessible residues in the transmembrane beta-barrel of OmpX.
European biophysics journal : EBJ (2009-07-30)
Laurent J Catoire, Manuela Zoonens, Carine van Heijenoort, Fabrice Giusti, Eric Guittet, Jean-Luc Popot
ABSTRACT
The atomic structure of OmpX, the smallest member of the bacterial outer membrane protein family, has been previously established by X-ray crystallography and NMR spectroscopy. In apparent conflict with electrophysiological studies, the lumen of its transmembrane beta-barrel appears too tightly packed with amino acid side chains to let any solute flow through. In the present study, high-resolution solution NMR spectra were obtained of OmpX kept water-soluble by either amphipol A8-35 or the detergent dihexanoylphosphatidylcholine. Hydrogen/deuterium exchange measurements performed after prolonged equilibration show that, whatever the surfactant used, some of the amide protons of the membrane-spanning region exchange much more readily than others, which likely reflects the dynamics of the barrel.