MilliporeSigma
  • Pyrethroid and organophosphate insecticide exposure in the 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine mouse model of Parkinson's disease: an immunohistochemical analysis of tyrosine hydroxylase and glial fibrillary acidic protein in dorsolateral striatum.
Pyrethroid and organophosphate insecticide exposure in the 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine mouse model of Parkinson's disease: an immunohistochemical analysis of tyrosine hydroxylase and glial fibrillary acidic protein in dorsolateral striatum.
Toxicology and industrial health (2009-03-26)
C A Dodd, B G Klein
ABSTRACT
The pyrethroid insecticide permethrin and the organophosphate insecticide chlorpyrifos can experimentally produce Parkinson's disease (PD)-associated changes in the dopaminergic nigrostriatal pathway, short of frank degeneration, although at doses considerably higher than from a likely environmental exposure. The ability of permethrin (200 mg/kg), chlorpyrifos (50 mg/kg), or combined permethrin + chlorpyrifos to facilitate nigrostriatal damage in the 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) (30 mg/kg) C57BL/6 mouse model of PD was investigated in three separate experiments. Tyrosine hydroxylase (TH) and glial fibrillary acidic protein (GFAP) immunohistochemistry assessed nigrostriatal degeneration or nigrostriatal damage more subtle than frank degeneration. Four fields in the dorsolateral caudate-putamen were examined at two rostrocaudal locations. The dopaminergic neurotoxin MPTP decreased striatal TH immunopositive neuropil and increased GFAP immunopositive neuropil. Neither permethrin nor chlorpyrifos, alone or in combination, altered the effects of MPTP upon TH or GFAP immunostaining. Permethrin alone increased striatal GFAP immunopositive neuropil but not when combined with chlorpyrifos treatment. Therefore, combined administration of the two insecticides appeared to protect against an increase in a neuropathological indicator of striatal damage seen with permethrin treatment alone. Differences compared with analysis of entire striatum emphasize the value of varying the topographic focus used to assess nigrostriatal degeneration in studies of insecticides in PD.
Sigma-Aldrich
4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, technical grade
Pricing and availability is not currently available.