Merck
  • One-pot conversion of thebaine to hydrocodone and synthesis of neopinone ketal.
One-pot conversion of thebaine to hydrocodone and synthesis of neopinone ketal.
The Journal of organic chemistry (2008-12-17)
Robert J Carroll, Hannes Leisch, Lena Rochon, Tomas Hudlicky, D Phillip Cox
ABSTRACT
The ethylene glycol ketal of neopinone was prepared in a one-pot procedure by the reaction of thebaine with ethylene glyocol in the presence of p-toluenesulfonic acid. The ketal is also an intermediate in the conversion of thebaine to hydrocodone with ethylene glycol and Pd(OAc)(2), followed by hydrogenation. Additionally, a one-pot procedure for the conversion of thebaine to hydrocodone was achieved by employing palladium catalysis in aqueous medium. Palladium serves a dual purpose in this transformation, first for the activation of the dienol ether of thebaine and second as a hydrogenation catalyst. This procedure was found to be comparable to the two-step protocol which employs diimide reduction of thebaine followed by acid-catalyzed hydrolysis of the resulting 8,14-dihydrothebaine to hydrocodone. Experimental and spectral data are provided for all compounds.
Sigma-Aldrich
Thebaine, powder
Pricing and availability is not currently available.