MilliporeSigma
End of the line for cannabinoid receptor 1 as an anti-obesity target?
Nature reviews. Drug discovery (2008-12-02)
Dan Jones
ABSTRACT
A wave of terminations of development programmes for cannabinoid receptor 1 blockers for obesity indicates the demise of a drug class that was once anticipated to yield blockbusters. Nevertheless, lessons learned might help salvage something for future such approaches.
Sigma-Aldrich
(+)-Fenfluramine hydrochloride