MilliporeSigma
  • Comparison of potency of ephedrine and mephentermine for prevention of post-spinal hypotension in caesarean section.
Comparison of potency of ephedrine and mephentermine for prevention of post-spinal hypotension in caesarean section.
Anaesthesia and intensive care (2008-06-21)
M Mohta, D Agarwal, L K Gupta, A Tyagi, A Gupta, A K Sethi
ABSTRACT
The dosages and potency of intravenous mephentermine for prevention of post-spinal hypotension are not available in English literature. This study was designed to determine the minimum effective dose (ED50) of mephentermine and to compare its potency with that of ephedrine for prevention of post-spinal hypotension in parturients undergoing caesarean section. Dixon's up-down method of sequential allocation was used for vasopressor doses. Following administration of spinal anaesthesia, patients received a prophylactic infusion with 50 mg infused over a period of 30 minutes as the initial dose and dose intervals of 5 mg, of either ephedrine or mephentermine. The ED50 of ephedrine was 25.0 mg (95% CI 15.5 to 40.4 mg). For mephentermine, the up-down method was abandoned due to the success of the minimum dose possible but the ED50 appeared to be less than 5 mg. In conclusion, the minimum effective dose of mephentermine is much less than that of ephedrine for prevention of post-spinal hypotension. Another trial with a lower starting dose and smaller dose interval of mephentermine is required to determine the potency ratio of mephentermine and ephedrine.