MilliporeSigma
[Lichenoid contact eczema caused by color film developer].
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete (1991-04-01)
J Schoel, W Tilgen, P J Frosch
ABSTRACT
We report on a single case of a lichen planus like contact dermatitis caused by colour developers. We emphasize similarities to lichen planus with regard to clinical, histopathological and immunohistological aspects.