MilliporeSigma
  • The imidazoline RX871024 induces death of proliferating insulin-secreting cells by activation of c-jun N-terminal kinase.
The imidazoline RX871024 induces death of proliferating insulin-secreting cells by activation of c-jun N-terminal kinase.
Cellular and molecular life sciences : CMLS (2008-02-19)
I I Zaitseva, J Størling, T Mandrup-Poulsen, P-O Berggren, S V Zaitsev
ABSTRACT
An insufficient number of insulin-producing beta-cells is a major cause of defective control of blood glucose in both type 1 and type 2 diabetes. The aim of this study was to clarify whether the insulinotropic imidazolines can affect the survival of highly proliferating insulin-secreting cells, here exemplified by the MIN6 cell line. Our data demonstrate that RX871024, but not efaroxan, triggered MIN6 cell death and potentiated death induced by a combination of the pro-inflammatory cytokines interleukin-1beta, interferon- gamma and tumor necrosis factor-alpha. These effects did not involve changes in nitric oxide production but correlated with stimulation of c-jun N-terminal kinase (JNK) activity and activation of caspases-1, -3, -8 and -9. Our results suggest that the imidazoline RX871024 causes death of highly proliferating insulin-secreting cells, putatively via augmentation of JNK activity, a finding that may impact on the possibility of using compounds of similar activity in the treatment of diabetes.
Sigma-Aldrich
Efaroxan hydrochloride, ≥98% (HPLC)