MilliporeSigma
  • Effect of notoginsenoside R1 on hepatic microcirculation disturbance induced by gut ischemia and reperfusion.
Effect of notoginsenoside R1 on hepatic microcirculation disturbance induced by gut ischemia and reperfusion.
World journal of gastroenterology (2008-01-08)
Wei-Xing Chen, Fang Wang, Yu-Ying Liu, Qing-Jiang Zeng, Kai Sun, Xin Xue, Xiang Li, Ji-Ying Yang, Li-Hua An, Bai-He Hu, Jin-Hui Yang, Chuan-She Wang, Zhi-Xin Li, Lian-Yi Liu, Yan Li, Jun Zheng, Fu-Long Liao, Dong Han, Jing-Yu Fan, Jing-Yan Han
ABSTRACT
To assess the effect of notoginsenoside R1 on hepatic microcirculatory disturbance induced by gut ischemia/reperfusion (I/R) in mice. The superior mesenteric artery (SMA) of C57/BL mice was ligated for 15 min to induce gut ischemia followed by 30-min reperfusion. In another set of experiments, R1 was continuously infused (10 mg/kg per hour) from 10 min before I/R until the end of the investigation to study the influence of R1 on hepatic microcirculatory disturbance induced by gut I/R. Hepatic microcirculation was observed by inverted microscopy, and the vascular diameter, red blood cell (RBC) velocity and sinusoid perfusion were estimated. Leukocyte rolling and adhesion were observed under a laser confocal microscope. Thirty and 60 min after reperfusion, lactate dehydrogenase (LDH), alanine aminotransferase (ALT) and aspartate transaminase (AST) in peripheral blood were determined. The expression of adhesion molecules CD11b/CD18 in neutrophils and tumor necrosis factor-alpha (TNF-alpha), interleukin-6 (IL-6) and monocyte chemotactic protein-1 (MCP-1) in plasma were evaluated by flow cytometry. E-selectin and intercellular adhesion molecule-1 (ICAM-1) in hepatic tissue were examined by immunofluorescence. After gut I/R, the diameters of terminal portal venules and central veins, RBC velocity and the number of perfused sinusoids were decreased, while the leukocyte rolling and adhesion, the expression of E-selectin in hepatic vessels and CD18 in neutrophils, IL-6, MCP-1, LDH, ALT and AST were increased. R1 treatment attenuated these alterations except for IL-6 and MCP-1. R1 prevents I/R-induced hepatic microcirculation disturbance and hepatocyte injury. The effect of R1 is related to its inhibition of leukocyte rolling and adhesion by inhibiting the expression of E-selectin in endothelium and CD18 in neutrophils.
Sigma-Aldrich
Notoginsenoside R1, ≥98% (HPLC)
Pricing and availability is not currently available.