MilliporeSigma
  • Effects of the ether phospholipid AMG-PC on mast cells are similar to that of the ether lipid AMG but different from that of the analogue hexadecylphosphocholine.
Effects of the ether phospholipid AMG-PC on mast cells are similar to that of the ether lipid AMG but different from that of the analogue hexadecylphosphocholine.
Immunopharmacology (1991-07-01)
N Grosman
ABSTRACT
The ether lipid AMG (1-O-hexadecyl-2-O-methylglycerol) has previously been shown to have both enhancing and inhibitory effects on histamine release from isolated rat mast cells. In addition, a synergistic interaction with the phorbol ester TPA indicated protein kinase C activation. In the present investigation the effects of the related ether phospholipid AMG-PC (1-O-hexadecyl-2-O-methylglycero-3-phosphocholine) and the analogue hexadecylphosphocholine (miltefosine (HPC) are compared with those of AMG. HPC had only inhibitory effects, in contrast to AMG-PC which acted similarly on histamine release induced by compound 48/80, but had dual influence on responses to antigen and the ionophore A23187 and mainly exhibited a synergistic interaction with combinations of the ionophore and TPA. The influence of AMG-PC differed from that of AMG in some aspects whereas the character of its synergistic interaction with A23187 resembled that of AMG, in particular after pre-incubation with TPA. The results show that HPC does not share all biological activities of the antineoplastic ether lipids. They indicate that the effects of AMG-PC under appropriate conditions may be due to activation of protein kinase C but do not permit a distinction between direct effects and indirect through metabolic conversion to AMG.