MilliporeSigma
  • Possibilities of column coupling electrophoresis provided with a fiber-based diode array detection in enantioselective analysis of drugs in pharmaceutical and clinical samples.
Possibilities of column coupling electrophoresis provided with a fiber-based diode array detection in enantioselective analysis of drugs in pharmaceutical and clinical samples.
Journal of chromatography. A (2007-10-02)
Peter Mikus, Katarína Maráková, Jozef Marák, Dusan Kaniansky, Iva Valásková, Emil Havránek
ABSTRACT
The present work illustrated possibilities of column coupling electrophoresis combined with ionizable chiral selector and diode array detection (DAD) for the enantioselective analysis of trace drugs (pheniramine and its analogs) in pharmaceutical and clinical samples. Isotachophoresis (ITP), on-line coupled with capillary zone electrophoresis (CZE), served as an ideal injection technique (high sample load capacity, narrow and sharp drugs zones) of on-line pretreated samples (preseparation, purification and preconcentration of drugs) for the CZE stage. Enhanced (enantio)separation selectivity of CZE with ionizable chiral selector (carboxyethyl-beta-cyclodextrin recognized between drugs enantiomers on one hand as well as between drugs and sample matrix constituents on the other hand) enabled to obtain pure zones of the drugs enantiomers, suitable for their detection and quantitation. DAD in comparison with single wavelength UV detection enhanced value of analytical information verifying purity of drugs enantiomers zones (indicating interferents with different spectra to those of drugs). Obtained results indicated pure zones of interest confirming effective ITP-CZE (enantio)separation process. Distinguishing the trace analytes signals superposed on the baseline noise was provided with sufficient reliability (for this purpose the background correction and smoothing procedure had to be applied to the raw DAD spectra). The proposed ITP-CZE-DAD methods were characterized by favorable performance parameters (sensitivity, linearity, precision, recovery, accuracy, robustness, selectivity) and successfully applied for (i) enantiomeric purity testing of dexbrompheniramine in commercial pharmaceutical tablets and (ii) enantioselective metabolic study of pheniramine in human urine.
Product Description
Supelco
Brompheniramine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®