MilliporeSigma
  • Subcutaneous, intrathecal and periaqueductal grey administration of asimadoline and ICI-204448 reduces tactile allodynia in the rat.
Subcutaneous, intrathecal and periaqueductal grey administration of asimadoline and ICI-204448 reduces tactile allodynia in the rat.
European journal of pharmacology (2007-07-24)
Nadia L Caram-Salas, Gerardo Reyes-García, Gerd D Bartoszyk, Claudia I Araiza-Saldaña, Mónica Ambriz-Tututi, Héctor I Rocha-González, Rosaura Arreola-Espino, Silvia L Cruz, Vinicio Granados-Soto
ABSTRACT
The purpose of this study was to assess the possible antiallodynic effect of asimadoline ([N-methyl-N-[1S)-1-phenyl)-2-(13S))-3-hydroxypyrrolidine-1-yl)-ethyl]-2,2-diphenylacetamide HCl]) and ICI-20448 ([2-[3-(1-(3,4-Dichlorophenyl-N-methylacetamido)-2-pyrrolidinoethyl)-phenoxy]acetic acid HCl]), two peripheral selective kappa opioid receptor agonists, after subcutaneous, spinal and periaqueductal grey administration to neuropathic rats. Twelve days after spinal nerve ligation tactile allodynia was observed, along with an increase in kappa opioid receptor mRNA expression in dorsal root ganglion and dorsal horn spinal cord. A non-significant increase in periaqueductal grey was also seen. Subcutaneous (s.c.) administration of asimadoline and ICI-204448 (1-30 mg/kg) dose-dependently reduced tactile allodynia. This effect was partially blocked by s.c., but not intrathecal, naloxone. Moreover, intrathecal administration of asimadoline or ICI-204448 (1-30 mug) reduced tactile allodynia in a dose-dependent manner and this effect was completely blocked by intrathecal naloxone. Microinjection of both kappa opioid receptor agonists (3-30 mug) into periaqueductal grey also produced a naloxone-sensitive antiallodynic effect in rats. Our results indicate that systemic, intrathecal and periaqueductal grey administration of asimadoline and ICI-204448 reduces tactile allodynia. This effect may be a consequence of an increase in kappa opioid receptor mRNA expression in dorsal root ganglion, dorsal horn spinal cord and, to some extent, in periaqueductal grey. Finally, our data suggest that these drugs could be useful to treat neuropathic pain in human beings.