MilliporeSigma
Recent paramethoxymethamphetamine (PMMA) deaths in Taiwan.
Journal of analytical toxicology (2007-06-01)
Dong-Liang Lin, Hsiu-Chuan Liu, Hsin-Ling Yin
ABSTRACT
Paramethoxyamphetamine (PMA) and paramethoxymethamphetamine (PMMA) are methoxylated phenylethylamine derivatives that have been banned in Taiwan since December 2005. The case history and pathological and toxicological findings of eight recent PMMA fatalities were investigated. All specimens from these cases were initially identified by an AxSYM fluorescence polarization immunoassay screening test for amphetamines with a 300 ng/mL cutoff. Specimens screened positive were confirmed and quantitated by gas chromatography-mass spectrometry. The mean age of these PMMA-related fatalities was 18.9 +/- 4.4 years in the range of 14-25. Seven (87.5%) of these eight cases were men. The mean, standard deviation, and range of PMA found in the heart blood collected from these 8 cases were 0.213, 0.144, and 0.079-0.489 microg/mL, respectively. The corresponding data for PMMA were 4.312, 4.806, and 1.208-15.824 microg/mL, respectively. Other drugs, such as MDA, MDMA, ketamine, norketamine, hydroxymidazolam, methamphetamine, and pentobarbital, were also found in these cases.
Product Description
Supelco
PMA hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®