MilliporeSigma
  • On-demand generation of an efficient catalyst for pyridine formation from unactivated nitriles and alpha,omega-diynes using CoCl2-6H2O, dppe, and Zn.
On-demand generation of an efficient catalyst for pyridine formation from unactivated nitriles and alpha,omega-diynes using CoCl2-6H2O, dppe, and Zn.
Organic letters (2007-01-31)
Kouki Kase, Avijit Goswami, Kazuhiko Ohtaki, Emi Tanabe, Naoko Saino, Sentaro Okamoto
ABSTRACT
[reaction: see text] A highly practical, general method for catalytic formation of substituted pyridines from a variety of unactivated nitriles and alpha,omega-diynes is given. The reactions which were catalyzed by 5 mol % of dppe/CoCl2-6H2O in the presence of Zn powder (10 mol %) could proceed at rt to approximately 50 degrees C with high functional compatibility and regioselectivity.
Sigma-Aldrich
DPPE hydrochloride, ≥98% (HPLC)