MilliporeSigma
  • Involvement of Tetrahymena pyriformis and selected fungi in the elimination of anthracene, and toxicity assessment of the biotransformation products.
Involvement of Tetrahymena pyriformis and selected fungi in the elimination of anthracene, and toxicity assessment of the biotransformation products.
Ecotoxicology and environmental safety (2007-01-30)
P Guiraud, J L Bonnet, A Boumendjel, M Kadri-Dakir, M Dusser, J Bohatier, R Steiman
ABSTRACT
Anthracene (AC) is a non-mutagenic and non-carcinogenic, low-molecular-weight polycyclic aromatic hydrocarbon present in the environment. Its toxicity can be dramatically increased after solar-light exposure. Biotransformation capacities of AC by Tetrahymena pyriformis and a selection of eight micromycetes were studied, and the ability of these microorganisms to detoxify the polluted ecosystems was assessed. We showed that T. pyriformis was able to accumulate high amounts of AC without any transformation. In contrast, the fungi Cunninghamella elegans, Absidia fusca, Absidia cylindrospora, Rhodotorula glutinis, and Aspergillus terreus were able to transform AC with a high efficiency. Cytotoxicity assays conducted on HeLa cells and T. pyriformis showed that crude extract from A. fusca culture medium obtained after AC biotransformation was not toxic. For A. fusca and A. cylindrospora, 1-4 dihydroxyanthraquinone was shown to be the major product during the biotransformation process. This compound seemed to be a dead-end metabolite at least for the Absidia strains. The cytotoxicity of 1-4 dihydroxyanthraquinone was higher than that of AC to T. pyriformis but lower to HeLa cells. On the whole our results showed that the microorganisms studied were all able to decontaminate an AC-polluted ecosystem, either by accumulating or transforming the compound. A possible detoxification process resulting from AC biotransformation can be considered only using the human cell model.