MilliporeSigma
Onychoprotothecosis: report of the first case in Brazil.
International journal of dermatology (2006-09-12)
Clarisse Zaitz, Andrea Miranda Godoy, Valéria Maria de Sousa, Ligia Rangel B Ruiz, Andréa S Masada, Márcia Valéria Nobre, Ana Regina Alencar Santos, Ana Cristina Marques, Laura Hitomi Muramatu, Giovanna Letícia Hernandez Arrigada, Elisabeth Maria Heins-Vaccari, José Eduardo Costa Martins
ABSTRACT
The present report describes a case of onychoprotothecosis caused by Prototheca wickerhamii in a nonimmunocompromized female. Dermatological examination showed yellowish discoloration of the left and right forefinger nails, showing onicolysis and hyperkeratosis. The repeated isolation of the algae Prototheca (organisms morula-like) as well as the repeated culture in media, Sabouraud agar, confirmed the diagnosis. The patient was successfully treated with tioconazol 1% topic solution. The aim of this paper was to present a rare condition.
Sigma-Aldrich
Tioconazole, 97%