MilliporeSigma
  • Chromium(III) complexes of D-glucosaminic acid and their effect on decreasing blood sugar in vivo.
Chromium(III) complexes of D-glucosaminic acid and their effect on decreasing blood sugar in vivo.
Archiv der Pharmazie (2006-08-31)
Sheng-Ju Ou, Guang Chen, Zhi-Hua Lin, Zhi-Ping Bai, Chun-Ying Duan, Cai-Ping Mao
ABSTRACT
Two chromium(III) complexes of glucosaminic acid were synthesized by neutralization and exchange reaction. The formation of 1 : 1 and 2 : 3 (Cr : glucosaminate) complexes was confirmed by elemental analyses and spectroscopic studies. The effect of the complexes on decreasing blood sugar was investigated on type-2 diabetes model rats induced by tetraoxypyrimidine. The results indicated that the effect on decreasing blood sugar was comparable to that of picolinate chromium complex (Cr(pic)(3)) currently used world wide.
Sigma-Aldrich
D-Glucosaminic acid, ≥98% (TLC)