MilliporeSigma
  • Microwave-assisted synthesis of thieno[2,3-c]pyridine derivatives as a new series of allosteric enhancers at the adenosine A(1) receptor.
Microwave-assisted synthesis of thieno[2,3-c]pyridine derivatives as a new series of allosteric enhancers at the adenosine A(1) receptor.
Bioorganic & medicinal chemistry letters (2006-08-26)
Romeo Romagnoli, Pier Giovanni Baraldi, Allan R Moorman, Maria Antonietta Iaconinoto, Maria Dora Carrion, Carlota Lopez Cara, Mojgan Aghazadeh Tabrizi, Delia Preti, Francesca Fruttarolo, Stephen P Baker, Katia Varani, Pier Andrea Borea
ABSTRACT
The microwave-assisted aromatization method has been used for the synthesis of a series of novel thieno[2,3-c]pyridines. This rapid method produces compounds in good yield within minutes in comparison with conventional heating method. The synthesized molecules have been evaluated as a potential new series of allosteric enhancers acting at the adenosine A(1) receptor. In a functional assay, one compound (3h) showed activity comparable with that of reference compound PD 81,723.