MilliporeSigma
  • Selective and sensitized spectrophotometric determination of trace amounts of Ni(II) ion using alpha-benzyl dioxime in surfactant media.
Selective and sensitized spectrophotometric determination of trace amounts of Ni(II) ion using alpha-benzyl dioxime in surfactant media.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-07-18)
Mehrorang Ghaedi
ABSTRACT
Highly sensitive and interference-free sensitized spectrophotometric method for the determination of Ni(II) ions is described. The method is based on the reaction between Ni(II) ion and benzyl dioxime in micellar media in the presence of sodium dodecyl sulfate (SDS). The absorbance is linear from 0.1 up to 25.0 microg mL-1 in aqueous solution with repeatability (RSD) of 1.0% at a concentration of 1 microg mL-1 and a detection limit of 0.12 ng mL-1 and molar absorption coefficient of 68,600L mol-1 cm-1. The influence of reaction variables including type and amount of surfactant, pH, and amount of ligand and complexation time and the effect of interfering ions are investigated. The proposed procedure was applied to the determination of trace amounts of Ni(II) ion in tap water, river water, chocolate and vegetable without separation or organic solvent extraction.
Sigma-Aldrich
anti-Diphenylglyoxime, 97%
Pricing and availability is not currently available.