MilliporeSigma
  • Biochemical effects of nonylphenol polyethoxylate adjuvant, Diquat herbicide and their mixture on the three-spined stickleback (Gasterosteus aculeatus L.).
Biochemical effects of nonylphenol polyethoxylate adjuvant, Diquat herbicide and their mixture on the three-spined stickleback (Gasterosteus aculeatus L.).
Marine environmental research (2006-05-19)
W Sanchez, O Palluel, L Lagadic, S Aït-Aïssa, J-M Porcher
ABSTRACT
This study examined the response of 7-ethoxyresorufine-O-deethylase, glutathione-S-transferase, glutathione peroxidase, glutathione content, level of thiobarbituric acid reactive compounds and circulating vitellogenin, in three-spined sticklebacks after 21 days of exposure to Diquat herbicide, commercial nonylphenol polyethoxylate adjuvant and mixture between Diquat and adjuvant. The results showed that adjuvant exerted more important oxidative effects than Diquat and that mixture effects were unlike to single additivity. This study argues for ecotoxicological risk assessment of adjuvants and mixtures of adjuvants and pesticides.
Product Description
Supelco
Metoxuron, PESTANAL®, analytical standard