MilliporeSigma
  • Tetrazole thioacetanilides: potent non-nucleoside inhibitors of WT HIV reverse transcriptase and its K103N mutant.
Tetrazole thioacetanilides: potent non-nucleoside inhibitors of WT HIV reverse transcriptase and its K103N mutant.
Bioorganic & medicinal chemistry letters (2006-03-01)
Ester Muraglia, Olaf D Kinzel, Ralph Laufer, Michael D Miller, Gregory Moyer, Vandna Munshi, Federica Orvieto, Maria Cecilia Palumbi, Giovanna Pescatore, Michael Rowley, Peter D Williams, Vincenzo Summa
ABSTRACT
A series of aryltetrazolylacetanilides was synthesized and evaluated as HIV-1 non-nucleoside reverse transcriptase inhibitors on wild-type virus and on the clinically relevant K103N mutant strain. Extensive SAR investigation led to potent compounds, with nanomolar activity on K103N, and orally bioavailable in rats.