MilliporeSigma
  • 5-Hydroxytryptamine stimulates corticosteroid-sensitive CRF release from cultured foetal hypothalamic cells. Role of protein kinases.
5-Hydroxytryptamine stimulates corticosteroid-sensitive CRF release from cultured foetal hypothalamic cells. Role of protein kinases.
Brain research (1992-03-06)
S B Hu, S L Lightman, L A Tannahill
ABSTRACT
5-Hydroxytryptamine (5-HT) has been shown to activate the hypothalamo-pituitary-adrenal axis, possibly by a direct action on hypothalamic CRF synthesis and release. In order to study the mechanisms involved in this effect, foetal hypothalamic cells were cultured and corticotropin-releasing factor-41 (CRF) release was measured by radioimmunoassay. 5-HT induced CRF release in a dose-dependent manner. Further studies were performed with a specific protein kinase C inhibitor, H-7 (1-(5-isoquinolinesulfonyl)-2-methyl-piperazine) and a specific cyclic adenosine monophosphate-dependent protein kinase inhibitor, IP-20. Basal release of CRF-41 from the cultured hypothalamic cells was unaffected by IP-20 and was only diminished at a high (50 microM) concentration of H-7. 5-HT stimulated-CRF release, however, was blocked by both H-7 and IP-20. Dexamethasone and aldosterone both caused a dose-dependent inhibition of 5-HT induced CRF release. These results demonstrate that CRF can be released from hypothalamic neurons in response to 5-HT through a protein kinase C and protein kinase A dependent mechanism and that 5-HT stimulated CRF release can be inhibited by dexamethasone and aldosterone.