MilliporeSigma
  • Indole protects tryptophan indole-lyase, but not tryptophan synthase, from inactivation by trifluoroalanine.
Indole protects tryptophan indole-lyase, but not tryptophan synthase, from inactivation by trifluoroalanine.
Archives of biochemistry and biophysics (1992-08-01)
R S Phillips, R K Dua
ABSTRACT
Trifluoroalanine is a mechanism-based inactivator of Escherichia coli tryptophan indole-lyase (tryptophanase) and E. coli tryptophan synthase (R. B. Silverman and R. H. Abeles, 1976, Biochemistry 15, 4718-4723). We have found that indole is able to prevent inactivation of tryptophan indole-lyase by trifluoroalanine. The protection of tryptophan indole-lyase by indole exhibits saturation kinetics, with a KD of 0.03 mM, which is comparable to the KI for inhibition of pyruvate ion formation (0.01 mM) and the Km for L-tryptophan synthesis. Fluoride electrode measurements indicate the formation of 28 mol of fluoride ion per mole of enzyme during inactivation of tryptophan indole-lyase, and 121 mol of fluoride ion are formed per mole of enzyme in the presence of 2 mM indole during the same incubation period. 19F NMR spectra of reaction mixtures of tryptophan indole-lyase and trifluoroalanine showed evidence only for fluoride ion formation, in either the absence or the presence of indole, and difluoropyruvic acid was not detected. The partition ratio, kcat/kinact, is estimated to be 9. Tryptophan indole-lyase in the presence of trifluoroalanine exhibits visible absorption peaks at 446 and 478 nm, which decay at the same rate as inactivation. However, in the presence of 1 mM indole and trifluoralanine, tryptophan indole-lyase exhibits a peak only at 420 nm, and the spectra show a gradual increase at 300-310 nm with incubation. In contrast, tryptophan synthase is not protected by indole from inactivation by trifluoroalanine, and the absorption peak at 408 nm for the tryptophan synthase-trifluoroalanine complex is unaffected by indole. These results demonstrate that inactivation of tryptophan indole-lyase occurs via a catalytically competent species, probably the beta,beta-difluoro-alpha-aminoacrylate intermediate, which can be partitioned from inactivation to products by a reactive aromatic nucleophile, indole.
Sigma-Aldrich
3,3,3-Trifluoro-DL-alanine, 98%