MilliporeSigma
  • Stereospecific addition of H-P bond to alkenes: a simple method for the preparation of (R(P))-phenylphosphinates.
Stereospecific addition of H-P bond to alkenes: a simple method for the preparation of (R(P))-phenylphosphinates.
The Journal of organic chemistry (2005-11-19)
Li-Biao Han, Chang-Qiu Zhao
ABSTRACT
[reaction: see text] A variety of functionalized optically pure (R(P))-alkylphenylphosphinates are readily prepared by stereospecific radical or base-catalyzed additions of the easily available (R(P))-menthyl phenylphosphinate to alkenes.
Sigma-Aldrich
Phenylphosphinic acid, 99%