MilliporeSigma
  • The methyl-5 alpha-dihydrotestosterones mesterolone and drostanolone; gas chromatographic/mass spectrometric characterization of the urinary metabolites.
The methyl-5 alpha-dihydrotestosterones mesterolone and drostanolone; gas chromatographic/mass spectrometric characterization of the urinary metabolites.
The Journal of steroid biochemistry and molecular biology (1992-05-01)
D de Boer, E G de Jong, R A Maes, J M van Rossum
ABSTRACT
Before including the detection of the methyl-5 alpha-dihydrotestosterones mesterolone (1 alpha-methyl-17 beta-hydroxy-5 alpha-androstan-3-one) and drostanolone (2 alpha-methyl-17 beta-hydroxy-5 alpha-androstan-3-one) in doping control procedures, their urinary metabolites were characterized by gas chromatography/mass spectrometry. Several metabolites were found after enzymatic hydrolysis and conversion of the respective metabolites to their trimethylsilyl-enol-trimethylsilyl ether derivatives. The major metabolites of mesterolone and drostanolone were identified as 1 alpha-methyl-androsterone and 2 alpha-methyl-androsterone, respectively. The parent compounds and the intermediate 3 alpha,17 beta-dihydroxysteroid metabolites were detected as well. The reduction into the corresponding 3 beta-hydroxysteroids was a minor metabolic pathway. All metabolites were found to be conjugated to glucuronic acid.
Product Description
Supelco
Mesterolone, analytical standard
Pricing and availability is not currently available.