MilliporeSigma
  • Synthesis and properties of different metal complexes of the siderophore desferriferricrocin.
Synthesis and properties of different metal complexes of the siderophore desferriferricrocin.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2005-05-04)
Guozhang Zou, Gregory L Boyer
ABSTRACT
Desferriferricrocin is a cyclic hexa-peptide siderophore with three hydroxamates as primary coordination groups. It forms metal complexes with Fe(III), Cr(III), Al(III), Ga(III), Cu(II), and Zn(II). These complexes were prepared and characterized using UV-vis, circular dichroism spectroscopy (CD), nuclear magnetic resonance spectroscopy (NMR), and electrospray ionization mass spectroscopy (ESI-MS). The mononuclear trivalent metal complexes of desferriferricrocin were stable in aqueous solutions, and their coordination centers primarily adopted the lambda configuration. The formation of multinuclear complexes of desferriferricrocin was determined by ESI-MS. Desferriferricrocin was able to bind up to three Cu(II) and two Zn(II) respectively. Heteronuclear complexes containing one trivalent and one divalent were also determined. In these complexes, amide nitrogens were utilized as alternative binding groups of desferriferricrocin in addition to the primary binding groups, the hydroxamates.
Sigma-Aldrich
Ferrichrome Iron-free from Ustilago sphaerogena
Pricing and availability is not currently available.