MilliporeSigma
  • Role of serotonin (5-HT)2 receptors in cocaine self-administration and seeking behavior in rats.
Role of serotonin (5-HT)2 receptors in cocaine self-administration and seeking behavior in rats.
Pharmacological reports : PR (2005-04-26)
Małgorzata Filip
ABSTRACT
Previous studies have indicated a role of serotonin (5-HT)2 receptors in modulation of the behavioral effects of cocaine. In the present study, the efficacy of SR 46349B (a 5-HT(2A) receptor antagonist) or SDZ SER-082 (a 5-HT(2C) receptor antagonist) in altering cocaine seeking behavior was examined in rats. Rats were trained to press a lever for cocaine (0.5 mg/kg/infusion, iv) paired with the cue (light + tone). After stabilization of self-administration response, the animals underwent daily extinction sessions during which responding had no consequences. The cocaine seeking behavior was reinstated by cocaine priming (10 mg/kg, ip) or by presentation of the cue. Neither SR 46349B (0.25-1 mg/kg) nor SDZ SER-082 (0.25-1 mg/kg) altered the maintenance of cocaine self-administration. SR 46349B (0.5-1 mg/kg) decreased responding to the cocaine priming dose and reduced cue-induced reinstatement, while SDZ SER-082 failed to alter both cue- and cocaine priming-induced reinstatement. These findings indicate that 5-HT(2A) and 5-HT(2C) receptors are not significant to cocaine rewarding effects. However, they show the importance of the 5-HT(2A) receptors (but not 5-HT(2C) receptors) in cocaine-priming- and cue-provoked reinstatement. Since drugs that reduce cocaine seeking also alleviate cocaine craving, 5-HT(2A) receptor antagonists may be considered to be of possible clinical application for the treatment of cocaine dependence.
Sigma-Aldrich
SR-46349 hemifumarate salt, ≥98% (HPLC)