MilliporeSigma
  • A "locked-on," constitutively active mutant of the adenosine A1 receptor.
A "locked-on," constitutively active mutant of the adenosine A1 receptor.
European journal of pharmacology (2005-03-03)
Rianne A F de Ligt, Scott A Rivkees, Anna Lorenzen, Rob Leurs, Ad P IJzerman
ABSTRACT
We studied the wild-type human adenosine A1 receptor and three mutant receptors, in which the glycine at position 14 had been changed into an alanine, a leucine, or a threonine residue. All receptors were characterized in radioligand binding experiments, the wild-type and the Gly14Thr mutant receptor in greater detail. Both receptors were allosterically modulated by sodium ions and PD81,723 (2-amino-4,5-dimethyl-3-thienyl-[3(trifluoromethyl)-phenyl]methanone), although in a different way. All mutant receptors appeared to be spontaneously or "constitutively" active in a [35S]GTPgammaS binding assay, the first demonstration of the existence of such CAM (constitutively active mutant) receptors for the adenosine A1 receptor. The Gly14Thr mutant receptor was also constitutively active in another functional assay, i.e., the inhibition of forskolin-induced cAMP production in intact cells. Importantly, this mutant displayed a peculiar "locked-on" phenotype, i.e., neither agonist nor inverse agonist was capable of modulating the basal activity in both the GTPgammaS and the cAMP assay, unlike the wild-type and the two other mutant receptors.