MilliporeSigma
  • Differential inhibition by tedisamil (KC 8857) and glibenclamide of the responses to cromakalim and minoxidil sulphate in rat isolated aorta.
Differential inhibition by tedisamil (KC 8857) and glibenclamide of the responses to cromakalim and minoxidil sulphate in rat isolated aorta.
Naunyn-Schmiedeberg's archives of pharmacology (1992-02-01)
K Bray, U Quast
ABSTRACT
The effects of the K+ channel blockers tedisamil and glibenclamide on cromakalim- and minoxidil sulphate-induced 42K+ and 86Rb+ efflux and vasorelaxation in rat aorta, were investigated. In aortic strips preloaded with 42K+ or 86Rb+, cromakalim (1 mumol/l) induced increases in tracer efflux, which were concentration-dependently inhibited by tedisamil with similar potencies (pD2 approximately 7.3) but different amplitudes (maximum inhibition of 86Rb+ efflux to 0% of control, 42K+ efflux to 10 +/- 1%). The 42K+ efflux elicited by a low concentration of cromakalim (100 nmol/l) was, however, fully inhibited by tedisamil. The tracer effluxes induced by minoxidil sulphate were fully inhibited by tedisamil and glibenclamide (300 nM). Cromakalim and minoxidil sulphate, produced a concentration-dependent inhibition of noradrenaline (100 nmol/l)-induced tone, with pD2 values of approximately 7.3. Tedisamil (300 nmol/l) and glibenclamide (300 nmol/l), which inhibited cromakalim- and minoxidil sulphate-induced 42K+ and 86Rb+ efflux by greater than or equal to 80%, produced 2-fold and 40-fold shifts in the concentration-relaxation curve for cromakalim, and 3.5-fold and 2200-fold shifts in the concentration-relaxation curve for minoxidil sulphate, respectively. Similar shifts of the cromakalim concentration-relaxation curve in the presence of tedisamil and glibenclamide were also observed when the tissues were precontracted with potassium chloride (25 mmol/l). The results show that tedisamil and glibenclamide inhibit the cromakalim- and minoxidil sulphate-induced tracer effluxes with similar potencies whereas they differ greatly in their ability to inhibit the vasorelaxant effects of the two K+ channel openers.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Minoxidil Sulfate