Merck
  • [Flow injection chemiluminescent detection of acemetacin in KMnO4 - Na2 SO3 system].
[Flow injection chemiluminescent detection of acemetacin in KMnO4 - Na2 SO3 system].
Yao xue xue bao = Acta pharmaceutica Sinica (2005-02-09)
Zhi-Jie Zhang, Ya-Feng Zhuang, Huang-Xian Ju
ABSTRACT
To study the sensitizing effect of acemetacin (ACE) on the weak chemiluminescent (CL) reaction of KMnO4 with sulfite and establish a fast and convenient method for CL detection of ACE. Using the sensitizing effect of ACE on KMnO4-Na2SO3 system and flow injection technique to determine the concentration of ACE. Under optimal conditions, the CL intensity of 1.0 x 10(-2) mol x L(-1) H3PO4 - 5.0 x 10(-5) mol x L(-1) KMnO4 - 4.0 x 10(-4) mol x L(-1) Na2SO3 was proportional to the concentration of ACE ranging from 1.0 x 10(-7) to 1.0 x 10(-5) mol x L(-1). The detection limit of ACE was 6.9 x 10(-8) mol x L(-1) at 3sigma. Satisfactory results were obtained for determination of ACE at 2.5 x 10(-6) mol x L(-1). The present method showed good precision, high sensitivity and selectivity and could be used for fast and convenient detection of ACE. It would be of significance to the clinical and pharmacological study of acemetacin.