MilliporeSigma
Plasma membrane potential of neutrophils generated by the Na+ pump.
Biochimica et biophysica acta (1985-07-11)
C L Bashford, C A Pasternak
ABSTRACT
The plasma membrane potential of human neutrophils was monitored using the anionic dye oxonol-V. The cells maintain a potential of -75 +/- 17 mV when suspended in physiological saline solutions. The cells are scarcely depolarized by extracellular K+ and the depolarization induced by the chemotactic peptide fMet-Leu-Phe is of similar magnitude for cells suspended in 5 or 155 mM K+. Neutrophils are, however, depolarized by suspension in K+-free media or after treatment with ouabain. Neutrophils catalyse Na+-H+ exchange and possess other electroneutral ion transport systems. We propose that the neutrophil membrane potential is generated by an electrogenic Na+ pump, that osmotic stability is achieved by electroneutral ion transport systems and that electrical stability is maintained by anion leakage. Similar mechanisms may also operate in other biological membranes.