MilliporeSigma
  • Protic acid (HClO4 supported on silica gel)-mediated synthesis of 2,3-unsaturated-O-glucosides and a chiral furan diol from 2,3-glycals.
Protic acid (HClO4 supported on silica gel)-mediated synthesis of 2,3-unsaturated-O-glucosides and a chiral furan diol from 2,3-glycals.
The Journal of organic chemistry (2004-09-18)
Aditi Agarwal, Shikha Rani, Yashwant D Vankar
ABSTRACT
Perchloric acid supported on silica gel acts as an excellent reagent system in converting glucals into 2,3-unsaturated-O-glucosides in good to excellent yields in short reaction time with good alpha selectivity. Primary, secondary, and allylic alcohols, phenols, and thiols react with 3,4,6-tri-O-acetyl glucal with equal ease. In addition to this, a chiral furan diol is obtained from unprotected D-glucal or D-galactal in good yields.