MilliporeSigma
  • Degradation of estradiol and ethinyl estradiol by activated sludge and by a defined mixed culture.
Degradation of estradiol and ethinyl estradiol by activated sludge and by a defined mixed culture.
Applied microbiology and biotechnology (2004-08-04)
Stefanie Weber, Prisca Leuschner, Peter Kämpfer, Wolfgang Dott, Juliane Hollender
ABSTRACT
The aerobic degradation of the natural hormone 17-beta-estradiol (E2) and the synthetic hormone 17-alpha-ethinyl estradiol (EE2) was investigated in batch experiments with activated sludge from a conventional and a membrane sewage treatment plant. E2 was converted to estrone (E1), the well known metabolite, and further completely transformed within 3 days. The turnover rates of E2 did not differ greatly between conventional and membrane activated sludge. EE2 was persistent in both sludges. By several transfers into fresh E2-medium an enrichment culture could be selected that used E2 as growth substrate. Further enrichment and isolation led to a defined mixed culture consisting of two strains, which were identified by a polyphasic approach as Achromobacter xylosoxidans and Ralstonia sp., respectively. The culture used E2 and E1 as growth substrates and transformed estriol (E3) and 16-alpha-hydroxyestrone but not the xenoestrogens bisphenol A, alpha-zearalenol, mestranol or EE2. The turnover rates of E2 were 0.025-0.1 microg h(-1) cfu(-1) and did not depend on the steroid concentration.
Sigma-Aldrich
Mestranol, 99%