MilliporeSigma
  • Research shows 'poppers' use in connected with infection among MSM. Little is done to address problem.
Research shows 'poppers' use in connected with infection among MSM. Little is done to address problem.
AIDS alert (2004-07-27)
ABSTRACT
Researchers and experts on the use of inhaled nitrates among men who have sex with men (MSM) say clinicians, public health officials, and AIDS groups are ignoring the impact of poppers, a popular party inhalant, on HIV risk behaviors and even seroconversion.