MilliporeSigma
  • Stimulation-enhanced 3-O-methylglucose efflux from the frog sartorius: kinetics and properties of the system.
Stimulation-enhanced 3-O-methylglucose efflux from the frog sartorius: kinetics and properties of the system.
Biochimica et biophysica acta (1992-08-24)
G W Booz, C P Bianchi
ABSTRACT
The characteristics of the process by which contraction enhances glucose transport in the frog sartorius were studied. Electrical stimulation increased the permeability of muscles to 3-O-methylglucose (3-O-MeGlc), a nonmetabolizable glucose analogue, increasing efflux as well as uptake. Enhanced efflux was due to an increase in Vmax of the efflux process. A lactacidosis had no effect on basal 3-O-MeGlc efflux, and replacement of media Na+ with Li+ did not affect stimulation-induced uptake. Also, basal and stimulated uptake was not affected by 1 microM 12-O-tetradecanoylphorbol-13-acetate (TPA), a protein kinase C activator. Lastly, N-carbobenzoxy-glycyl-L-phenylalaninamide, which inhibits insulin-enhanced, but not basal, glucose uptake in adipocytes, inhibited both basal and stimulated 3-O-MeGlc fluxes in the frog sartorius. From these findings, we conclude: (1) contraction and exercise enhance glucose transport in muscle by increasing the number of transporters in the plasma membrane, or their turnover, by an unknown process; and (2) basal glucose transport of muscle, unlike that of adipocytes, can not be distinguished from stimulated transport on the basis of its insensitivity to N-carbobenzoxyglycyl-L-phenylalaninamide.