MilliporeSigma
  • [Chemical components of essential oils from the herb of Ligularia virgaurea].
[Chemical components of essential oils from the herb of Ligularia virgaurea].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2004-05-14)
Yan-li Tang, Yan-ru Deng, Han-qing Wang
ABSTRACT
To provide the foundation for reasonable utilization by analyzing the essential oils of Ligularia virgaurea. The essential oils were extracted by using steam distillation and separated with GC capillary columns. The components were quantitatively determined with normalization method, and were identified with GC-MS. 41 components were identified, which took up 72.73% of the essential oils. The main components of essential oils were 4-methyl-1-(1-methylethyl)-3-cyclohexen-1-ol(14.369%), crotonic acid, 2,2-dimethyl-butanoic acid, 1-methyl-3-(1-methylethyl)-benzene, (1s-endo)-1,7,7-trimethyl-bicyclo[2,2,1]heptan-2-ol, trans-1-methyl-4-(1-methylethyl)-2-cyclohexen-1-ol, alpha-cadinol and alpha,alpha,4-trimethyl-3-cyclohexene-1-methanol.
Sigma-Aldrich
2,2-Dimethylbutyric acid, 96%