MilliporeSigma
  • Design of N-acetyl-6-sulfo-beta-d-glucosaminide-based inhibitors of influenza virus sialidase.
Design of N-acetyl-6-sulfo-beta-d-glucosaminide-based inhibitors of influenza virus sialidase.
Bioorganic & medicinal chemistry (2004-03-17)
Kenji Sasaki, Yoshihiro Nishida, Mikie Kambara, Hirotaka Uzawa, Tadanobu Takahashi, Takashi Suzuki, Yasuo Suzuki, Kazukiyo Kobayashi
ABSTRACT
Biological activity of N-acetyl-6-sulfo-beta-d-glucosaminides (6-sulfo-GlcNAc 1) having a structural homology to N-acetylneuraminic acid (Neu5Ac 2) and 2-deoxy-2,3-dehydro-N-acetylneuraminic acid (Neu5Ac2en 3) was examined in terms of inhibitory activity against influenza virus sialidase (influenza, A/Memphis/1/71 H3N2). pNP 6-Sulfo-GlcNAc 1a was proved to show substantial activity to inhibit the virus sialidase (IC(50)=2.8 mM), though p-nitrophenyl (pNP) GlcNAc without 6-sulfo group and pNP 6-sulfo-GlcNH(3)(+) 1b without 2-NHAc showed little activity (IC(50) >50 mM). The activity was enhanced nearly 100-fold when the pNP group of 1a was converted to p-acetamidophenyl one 5 (IC(50)=30 microM) or replaced with 1-naphthyl 6 (IC(50)=10 microM) or n-propyl one 8 (IC(50)=11 microM).
Sigma-Aldrich
D-Glucosaminic acid, ≥98% (TLC)