MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Translocation of beta-catenin into the nucleus independent of interactions with FG-rich nucleoporins.
Translocation of beta-catenin into the nucleus independent of interactions with FG-rich nucleoporins.
Experimental cell research (2003-10-22)
Eun-Kyung Suh, Barry M Gumbiner
ABSTRACT
beta-Catenin nuclear import has been found to be independent of classical nuclear localization signal (NLS) nuclear import factors. Here, we test the hypothesis that beta-catenin interacts directly with nuclear pore proteins to mediate its own transport. We show that beta-catenin, unlike importin-beta, does not interact detectably with Phe/Gly(FG)-repeat-rich nuclear pore proteins or nucleoporins (Nups). Moreover, unlike NLS-containing proteins, beta-catenin nuclear import is not inhibited by wheat germ agglutinin (WGA) or excess importin-beta. These results suggest beta-catenin nuclear translocation does not involve direct interactions with FG-Nups. However, beta-catenin has two regions that can target it to the nucleus, and its import is cold sensitive, indicating that beta-catenin nuclear import is still an active process. Transport is blocked by a soluble form of the C-cadherin cytoplasmic domain, suggesting that masking of the nuclear targeting signal may be a mechanism of regulating beta-catenin subcellular localization.