MilliporeSigma
  • Synthesis of carbocyclic and acyclic nucleosides possessing 2-fluoroadenine derivatives and their inhibitory activities against Plasmodium falciparum SAH hydrolase.
Synthesis of carbocyclic and acyclic nucleosides possessing 2-fluoroadenine derivatives and their inhibitory activities against Plasmodium falciparum SAH hydrolase.
Nucleic acids research. Supplement (2001) (2003-09-27)
Yukio Kitade, Hiroharu Kojima, Fazila Zulfiqur, Saori Yabe, Daisuke Yamagiwa, Yasutomo Ito, Masayuki Nakanishi, Yoshihito Ueno, Hye-Sook Kim, Yusuke Wataya
ABSTRACT
Carbocyclic and acyclic nucleosides possessing 2-fluoroadenine, such as 2-fluoronoraristeromycin (6) and 2-fluoro-9-[(2S,3R)-2,3,4-trihydroxy-butyl-1-yl]adenine (8), were synthesized and their inhibitory activities against human and Plasmodium falciparum recombinant SAH hydrolase were investigated.
Sigma-Aldrich
2-Fluoroadenine, 96%