MilliporeSigma
  • Use of erythromycin for the treatment of severe chronic constipation in children.
Use of erythromycin for the treatment of severe chronic constipation in children.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas (2003-09-23)
M A Bellomo-Brandão, E F Collares, E A L da-Costa-Pinto
ABSTRACT
The efficacy of erythromycin was assessed in the treatment of 14 children aged 4 to 13 years with refractory chronic constipation, and presenting megarectum and fecal impaction. A double-blind, placebo- controlled, crossover study was conducted at the Pediatric Gastroenterology Outpatient Clinic of the University Hospital. The patients were randomized to receive placebo for 4 weeks followed by erythromycin estolate, 20 mg kg-1 day-1, divided into four oral doses for another 4 weeks, or vice versa. Patient outcome was assessed according to a clinical score from 12 (most severe clinical condition) to 0 (complete recovery). At enrollment in the study and on the occasion of follow-up medical visits at two-week intervals, patient score and laxative requirements were recorded. During the first 30 days, the mean SD clinical score for the erythromycin group (N = 6) decreased from 8.2+/-2.3 to 2.2+/-1.0 while the score for the placebo group (N = 8) decreased from 7.8+/-2.1 to 2.9+/-2.8. During the second crossover phase, the score for patients on erythromycin ranged from 2.9+/-2.8 to 2.4+/-2.1 and the score for the patients on placebo worsened from 2.2+/-1.0 to 4.3+/-2.3. There was a significant improvement in score when patients were on erythromycin (P < 0.01). Mean laxative requirement was lower when patients ingested erythromycin (P < 0.05). No erythromycin-related side effects occurred. Erythromycin was useful in this group of severely constipated children. A larger trial is needed to fully ascertain the prokinetic efficacy of this drug as an adjunct in the treatment of severe constipation in children.
Product Description
Erythromycin estolate, European Pharmacopoeia (EP) Reference Standard