MilliporeSigma
  • Isomer-specific analysis of chlorinated biphenyls, naphthalenes and dibenzofurans in Delor: polychlorinated biphenyl preparations from the former Czechoslovakia.
Isomer-specific analysis of chlorinated biphenyls, naphthalenes and dibenzofurans in Delor: polychlorinated biphenyl preparations from the former Czechoslovakia.
Environmental pollution (Barking, Essex : 1987) (2003-08-21)
Sachi Taniyasu, Kurunthachalam Kannan, Ivan Holoubek, Alena Ansorgova, Yuichi Horii, Nobuyasu Hanari, Nobuyoshi Yamashita, Kenneth M Aldous
ABSTRACT
Technical polychlorinated byphenyl (PCB) preparations--Delors 103, 104, 105, and 106--produced in the former Czechoslovakia were analyzed for their chlorobiphenyl (CB), chloronaphthalene (PCN) and chlorinated dibenzofuran (PCDF) composition and content using high resolution gas chromatography-mass spectrometry techniques. The congener patterns of Delors 103, 104, 105, and 106 resembled Aroclors 1242, 1248, 1254, and 1260. Delors contained PCNs and PCDFs, as impurities, at microgram per gram concentrations. Concentrations of PCNs and PCDFs in Delors were greater than those found in the corresponding Aroclors. The potential for the emissions of PCNs and PCDFs from Delor was estimated to be 3680 and 860 kg, respectively. Non- and mono-ortho PCBs were the major contributors to 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) equivalents in Delor mixtures.
Product Description
Supelco
PCB No 31, analytical standard