MilliporeSigma
[The effect of cyclosporin A on the islet cells of mice].
Biulleten' eksperimental'noi biologii i meditsiny (1992-11-01)
E D Gol'dberg, V A Eshchenko, V D Bovt, A Ia Tolok
ABSTRACT
The action of immunosuppressive agent ciclosporin A on the insular apparatus was studied with an original method of cytochemical zinc detection in experiments on mice. The drug was found to lower zinc levels in pancreatic beta-cells. In ciclosporin A pretreatment the sensitivity of mice to zinc-binding diabetogenic agent enhanced.