MilliporeSigma
[Pharmacological comparison between virosecurinine and securinine].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2003-02-18)
H Pu, J Zhao, B Pong, S Xu
ABSTRACT
Pharmacological activities of virosecurinine (Vse) and securinine (Sec) were studied. The results showed that acute toxicity of Vse was 1/13.6 that of Sec, and Vse had no convulsive effects on rats or frogs, while Sec had. The results also showed that Vse and Sec could elevate blood pressure and excite respiration in cats.
Sigma-Aldrich
Securinine, ≥98% (HPLC)