MilliporeSigma
  • Altretamine (hexamethylmelamine) in the treatment of platinum-resistant ovarian cancer: a phase II study.
Altretamine (hexamethylmelamine) in the treatment of platinum-resistant ovarian cancer: a phase II study.
Gynecologic oncology (2003-02-15)
Nina Keldsen, Hanne Havsteen, Ignace Vergote, Kamma Bertelsen, Anders Jakobsen
ABSTRACT
To evaluate the activity of oral Altretamine in women with epithelial ovarian carcinoma who responded (PR or CR) to first line chemotherapy but relapsed within 6 months. The protocol was later amended to include patients with relapse within 12 months. A multicentric phase II trial. The patients had to have measurable disease. No more than one prior chemotherapy regiment was allowed. The patients were treated with 260 mg/m(2)/day of Altretamine in four divided doses for 2 weeks, repeated every 4 weeks. The response was evaluated after every two courses. Thirty-one eligible patients were treated with a median of 3 courses of Altretamine (range 1-12). Hematological toxicity was minimal. Gastrointestinal toxicity was common. Response evaluation was possible for 26 patients. Three patients (9.7% intent-to-treat) achieved a partial response. Eight patients had stable disease, and 15 patients had progressive disease after two treatment courses. The median time to progression was 10 weeks (range, 5-51 weeks). Medial survival was 34 weeks (range, 7-112+). Altretamine should not be chosen as standard treatment in patients with platinum-resistant recurrent ovarian cancer. However, Altretamine represents a useful alternative in patients who prefer oral treatment or when socioeconomic considerations are an important issue.
Sigma-Aldrich
2,4,6-Tris(dimethylamino)-1,3,5-triazine, 96%