MilliporeSigma
  • Grapefruit extract activity in the control of rose powdery mildew and black spot.
Grapefruit extract activity in the control of rose powdery mildew and black spot.
Mededelingen (Rijksuniversiteit te Gent. Fakulteit van de Landbouwkundige en Toegepaste Biologische Wetenschappen) (2002-11-12)
A T Wojdyła
ABSTRACT
Efficacy of grapefruit extract (a.i. of Biosept 33 SL) in the control of Sphaerotheca pannosa var. rosae and Diplocarpon rosae on roses was investigated during 1998-1999. The extract was applied as plant spray in concentrations from 0.017 to 0.099%. First treatment of rose shrubs was done when visible disease symptoms occurred on leaves and spraying was repeated 3 (in plastic tunnel) or 10-times (in the field) at weekly intervals. In the second experiment roses with visible powdery mildew symptoms were sprayed once with grapefruit extract. Leaves were sampled one or 7 days after the extract application and germination of spores of S. pannosa var. rosae on potato dextrose agar was evaluated. In the next experiment roses grown under plastic tunnel were sprayed once with the tested preparation. After 24 hours leaves were collected and appearance of fungal hyphae and spores of S. pannosa var. rosae was studied in scanning electron microscope. In the control of S. pannosa var. rosae grapefruit extract at conc. 0.066% was as effective as triforine (standard) applied at 0.027%. Reduction of concentration resulted in the decreased efficacy of the tested preparation. Spores of S. pannosa var. rosae collected one day after grapefruit extract application germinated in about 5%. Analyses of spore vitality 6 days letter showed that only about 15% of conidia could germinated on PDA agar. In contrary, spores from untreated leaves germinated in about 95%. Scanning electrone microscope analysis of leaves taken from plants protected with grapefruit extract showed that most of hyphae were separated from leaf surface. Almost all hyphae and spores were degenerated. In the control of D. rosae the preparation in all tested concentrations gave satisfactory results but was less effective than triforine.
Product Description
Supelco
Triforin, PESTANAL®, analytical standard