MilliporeSigma
  • Erythro- and threo-2-hydroxynonyl substituted 2-phenyladenines and 2-phenyl-8-azaadenines: ligands for A1 adenosine receptors and adenosine deaminase.
Erythro- and threo-2-hydroxynonyl substituted 2-phenyladenines and 2-phenyl-8-azaadenines: ligands for A1 adenosine receptors and adenosine deaminase.
Farmaco (Societa chimica italiana : 1989) (2002-05-07)
Giuliana Biagi, Irene Giorgi, Oreste Livi, Federica Pacchini, Pietro Rum, Valerio Scartoni, Barbara Costa, Maria Rosa Mazzoni, Laura Giusti
ABSTRACT
erythro-2-Phenyl-9-(2-hydroxy-3-nonyl)adenine and its 8-aza analog were prepared and showed a very high inhibitory activity towards adenosine deaminase (ADA), with Ki 0.55 and 1.67 nM, respectively, and high affinity for A1 adenosine receptors, with Ki 28 and 2.8 nM, respectively. To increase affinity for A1 receptors we introduced a substituent on the N6 position such as alkyl or cycloalkyl groups, which are present in effective agonists or antagonists. Furthermore, for some compounds, we prepared the two diastereoisomers erythro and threo to verify whether the binding with A1 receptors is stereoselective, as in ADA. Results show that some of the synthesised compounds are good inhibitors for ADA and good ligands for A1, and the erythro diastereoisomers are more active than the threo ones. The experimental evidence allows us to hypothesise some similarity in the three dimensional structures of the binding site of the two proteins, ADA and A1 adenosine receptor, in spite of lacking any homologies in the amino acid sequences.