- [Study on the connection of alcohol motivation with zinc content changes in the hippocampus].
[Study on the connection of alcohol motivation with zinc content changes in the hippocampus].
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994) (2001-08-25)
V D Bovt, V A Ieshchenko, M M Mal'ko, O M Kuchkovs'kyĭ, N V Hryhorova
PMID11519251
ABSTRACT
Alcohol dependence development is accompanied by zinc content decrease in the hippocampus. Young age, repeated physical loads and immobilization are factors, that promote such changes development in more expressed degree.