MilliporeSigma
  • Inhibition of poly (ADP-ribose) synthetase by gene disruption or inhibition with 5-iodo-6-amino-1,2-benzopyrone protects mice from multiple-low-dose-streptozotocin-induced diabetes.
Inhibition of poly (ADP-ribose) synthetase by gene disruption or inhibition with 5-iodo-6-amino-1,2-benzopyrone protects mice from multiple-low-dose-streptozotocin-induced diabetes.
British journal of pharmacology (2001-07-17)
J G Mabley, W L Suarez-Pinzon, G Haskó, A L Salzman, A Rabinovitch, E Kun, C Szabó
ABSTRACT
Activation of poly(ADP-ribose) synthetase (PARS, also termed polyADP-ribose polymerase or PARP) has been proposed as a major mechanism contributing to beta-cell destruction in type I diabetes. In the present study, we have investigated the role of PARS in mediating the induction of diabetes and beta-cell death in the multiple-low-dose-streptozotocin (MLDS) model of type I diabetes. Mice genetically deficient in PARS were found to be less sensitive to MLDS than wild type mice, with a lower incidence of diabetes and reduced hyperglycemia. A potent inhibitor of PARS, 5-iodo-6-amino-1,2-benzopyrone (INH(2)BP), was also found to protect mice from MLDS and prevent beta-cell loss, in a dose-dependent manner. Paradoxically, in the PARS deficient mice, the compound increased the onset of diabetes. In vitro the cytokine combination; interleukin-1beta, tumor necrosis factor-alpha and interferon-gamma inhibited glucose-stimulated insulin secretion from isolated rat islets of Langerhans and decreased RIN-5F cell viability. The PARS inhibitor, INH(2)BP, protected both the rat islets and the beta-cell line, RIN-5F, from these cytokine-mediated effects. These protective effects were not mediated by inhibition of cytokine-induced nitric oxide formation. Inhibition of PARS by INH(2)BP was unable to protect rat islet cells from cytokine-mediated apoptosis. Cytokines, peroxynitrite and streptozotocin were all shown to induce PARS activation in RIN-5F cells, an effect suppressed by INH(2)BP. The present study provides evidence for in vivo PARS activation contributing to beta-cell damage and death in the MLDS model of diabetes, and indicates a role for PARS activation in cytokine-mediated depression of insulin secretion and cell viability in vitro.