MilliporeSigma
  • Anxiolytic-like actions of melatonin, 5-metoxytryptophol, 5-hydroxytryptophol and benzodiazepines on a conflict procedure.
Anxiolytic-like actions of melatonin, 5-metoxytryptophol, 5-hydroxytryptophol and benzodiazepines on a conflict procedure.
Progress in neuro-psychopharmacology & biological psychiatry (2000-02-05)
E B Naranjo-Rodriguez, A O Osornio, E Hernandez-Avitia, V Mendoza-Fernández, A Escobar
ABSTRACT
Male Wistar rats (120-230 g) were used in these experiments. Some rats were deprived of water for 48 h before testing in a conflict procedure. Then, after 20 licks from a water bottle with a metal drinking tube, the animal received an electric shock (0.5 mA/500 msec). The effects of two classical anxiolytic drugs, DIAZ (1.0, 2.0 and 4.0 mg/Kg b.wt), and CDP (5.0 and 10.0 mg/Kg b.wt) were compared to those produced by MEL (0.1, 0.2, 0.5, 1.0 and 2.0 mg/Kg b.wt), 5-MTOPHOL (1.0 and 2.0 mg/Kg b.wt), 5-HTOPHOL (1.0 and 2.0 mg/Kg b.wt) and vehicle solution. Anxiolytic drugs as well as MEL produced a dose-dependent increase in the number of shocks received. The results suggest that the three pineal indoles are involved in the modulation of the stress responses. MEL showed a higher potency than the other indoles.