MilliporeSigma
  • On-resin macrocyclization of peptides via intramolecular SnAr reactions.
On-resin macrocyclization of peptides via intramolecular SnAr reactions.
Bioorganic & medicinal chemistry letters (1999-08-28)
C Fotsch, G Kumaravel, S K Sharma, A D Wu, J S Gounarides, N R Nirmala, R C Petter
ABSTRACT
On-resin macrocyclization via an SNAr reaction is employed in the synthesis of tocinoic acid analogs. Specifically, an N-terminal nitrofluorobenzene is attacked by a nucleophilic C-terminal sidechain. The remaining nitro group can be reduced and acylated. NMR is used to compare the conformation of the new macrocyclic peptides to tocinoic acid.